Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gng7
Entrez Gene ID
14708
Gene Name
guanine nucleotide binding protein (G protein), gamma 7
Gene Aliases
AI840417
Chromosome Location
Chr.10: 80948624 - 81014961 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038655.1 NP_001033744.1 2 - 3 189 106
NM_010319.3 NP_034449.2 2 - 3 126 106
GenBank mRNA AK141541.1 - 2 - 3 98 106
AK147843.1 - 2 - 3 89 106
AK158963.1 - 2 - 3 189 106
AY035844.1 - 2 - 3 109 106
BC034108.1 - 1 - 2 53 106
BY271138.1 - 2 - 3 146 106

View Full Product Details