Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Reep6
Entrez Gene ID
70335
Gene Name
receptor accessory protein 6
Gene Aliases
0610011M24Rik, Dp1l1
Chromosome Location
Chr.10: 80330003 - 80336441 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204931.1 NP_001191860.1 4 - 5 603 85
GenBank mRNA AK002562.1 - 4 - 5 601 85
BC029741.1 - 4 - 5 590 85

View Full Product Details