Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trim29
Entrez Gene ID
72169
Gene Name
tripartite motif-containing 29
Gene Aliases
1110047J21Rik, 2810431N19Rik, 4732461M22Rik, AI119726
Chromosome Location
Chr.9: 43310715 - 43336125 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023655.2 NP_076144.2 4 - 5 1450 63
GenBank mRNA AF230390.1 - 2 - 3 323 63
AK013219.1 - 3 - 4 501 63
AK028448.1 - 4 - 5 683 63
BC006699.1 - 4 - 5 1181 63
BC027353.1 - 4 - 5 1365 63

View Full Product Details