The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_023655.2 | NP_076144.2 | 7 - 8 | 1739 | 77 |
GenBank mRNA | AK013219.1 | - | 6 - 7 | 790 | 77 |
AK028448.1 | - | 7 - 8 | 972 | 77 | |
BC006699.1 | - | 7 - 8 | 1470 | 77 | |
BC027353.1 | - | 7 - 8 | 1654 | 77 |