The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110306.1 | NP_001103776.1 | 1 - 2 | 427 | 112 |
GenBank mRNA | CJ179527.1 | - | 1 - 2 | 175 | 112 |