Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Egln2
- Entrez Gene ID
-
112406
- Gene Name
- egl-9 family hypoxia-inducible factor 2
- Gene Aliases
-
0610011A13Rik, C85656, Hif-p4h-1, Ier4, Phd1, SM-20
- Chromosome Location
-
Chr.7: 27158658 - 27166802 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK166661.1 |
- |
1 - 2 |
99 |
79 |
View Full Product Details