The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025837.2 | NP_080113.1 | 5 - 6 | 703 | 80 |
GenBank mRNA | AF244360.1 | - | 5 - 6 | 685 | 80 |
AK003289.1 | - | 3 - 4 | 463 | 80 | |
AK003896.1 | - | 5 - 6 | 704 | 80 | |
AK132777.1 | - | 5 - 6 | 704 | 80 | |
BC009068.1 | - | 5 - 6 | 681 | 80 | |
BC051163.1 | - | 5 - 6 | 681 | 80 |