Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Alg9
Entrez Gene ID
102580
Gene Name
asparagine-linked glycosylation 9 (alpha 1,2 mannosyltransferase)
Gene Aliases
8230402H15Rik, AI747665, B430313H07Rik, Dibd1
Chromosome Location
Chr.9: 50774881 - 50843639 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133981.2 NP_598742.1 10 - 11 1334 65
NR_045601.1 9 - 10 1263 65
GenBank mRNA AK050335.1 - 10 - 11 1255 65
AK054293.1 - 10 - 11 1240 65
AK086674.1 - 10 - 11 1274 65
AK149592.1 - 9 - 10 1185 65
BC021791.1 - 10 - 11 1229 65

View Full Product Details