Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aagab
Entrez Gene ID
66939
Gene Name
alpha- and gamma-adaptin binding protein
Gene Aliases
2310007F21Rik, p34
Chromosome Location
Chr.9: 63602629 - 63641897 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025857.2 NP_080133.1 3 - 4 396 69
GenBank mRNA AK017714.1 - 3 - 4 365 69
AK159897.1 - 3 - 4 397 69
AK209997.1 - 2 - 3 188 69
AK214885.1 - 2 - 3 188 69
AK215257.1 - 2 - 3 188 69
BC027311.1 - 3 - 4 392 69

View Full Product Details