The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001177629.1 | NP_001171100.1 | 14 - 15 | 1556 | 65 |
NM_010345.4 | NP_034475.2 | 15 - 16 | 1881 | 65 | |
GenBank mRNA | AK030727.1 | - | 15 - 16 | 1816 | 65 |
AK032927.1 | - | 15 - 16 | 1862 | 65 | |
AK163841.1 | - | 15 - 16 | 1862 | 65 | |
BC016111.1 | - | 14 - 15 | 1556 | 65 | |
BC053842.1 | - | 15 - 16 | 1848 | 65 |