Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc22a16
Entrez Gene ID
70840
Gene Name
solute carrier family 22 (organic cation transporter), member 16
Gene Aliases
4921504E14Rik, CT2, FLIPT2, OCT6, OKB1
Chromosome Location
Chr.10: 40570321 - 40604135 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027572.1 NP_081848.1 5 - 6 1418 148
GenBank mRNA AK014809.1 - 2 - 3 651 148
BC100473.1 - 5 - 6 1418 148
BC138326.1 - 5 - 6 1429 148
BC138327.1 - 5 - 6 1429 148
DQ164804.2 - 5 - 6 1444 148
DQ164805.1 - 5 - 6 1405 148

View Full Product Details