Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Txlna
Entrez Gene ID
109658
Gene Name
taxilin alpha
Gene Aliases
2600010N21Rik, AI425952, AW209241, Il14, Txln
Chromosome Location
Chr.4: 129626077 - 129641084 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005506.3 NP_001005506.2 1 - 2 548 72
NM_001199695.1 NP_001186624.1 2 - 3 265 72
GenBank mRNA AK078192.1 - 2 - 3 244 72
AK135290.1 - 1 - 2 396 72
AK141252.1 - 2 - 3 199 72
AK150240.1 - 2 - 3 222 72
AK160829.1 - 1 - 2 396 72
AK161649.1 - 2 - 3 228 72
AK168418.1 - 1 - 2 549 72
AK171101.1 - 2 - 3 241 72
BC060227.1 - 1 - 2 383 72
BC065115.1 - 1 - 2 383 72

View Full Product Details