Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cand2
Entrez Gene ID
67088
Gene Name
cullin-associated and neddylation-dissociated 2 (putative)
Gene Aliases
2210404G23Rik, Tp120b, mKIAA0667
Chromosome Location
Chr.6: 115774557 - 115805555 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025958.2 NP_080234.2 5 - 6 816 102
GenBank mRNA AK129186.1 - 5 - 6 787 102
AK145112.1 - 5 - 6 801 102
BC056365.1 - 5 - 6 798 102
BC058338.1 - 4 - 5 631 102

View Full Product Details