Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmcc1
Entrez Gene ID
330401
Gene Name
transmembrane and coiled coil domains 1
Gene Aliases
3632431M01Rik, mKIAA0779
Chromosome Location
Chr.6: 116018611 - 116193379 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177412.1 NP_803131.1 4 - 5 1805 93
GenBank mRNA AK028322.1 - 2 - 3 1434 93
AK154540.1 - 2 - 3 1491 93
AK173022.1 - 4 - 5 1794 93
BC029759.1 - 3 - 4 1627 93
BC157922.1 - 3 - 4 1728 93

View Full Product Details