Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Morc2a
Entrez Gene ID
74522
Gene Name
microrchidia 2A
Gene Aliases
8430403M08Rik, Zcwcc1
Chromosome Location
Chr.11: 3649494 - 3690477 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159288.1 NP_001152760.1 16 - 17 2222 123
NM_198162.1 NP_937805.1 15 - 16 1549 123
GenBank mRNA AK173042.1 - 16 - 17 2222 123
BC059243.1 - 15 - 16 1549 123
BC063082.1 - 4 - 5 489 123

View Full Product Details