Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxk1
Entrez Gene ID
17425
Gene Name
forkhead box K1
Gene Aliases
A630048H08Rik, AI463295, ENSMUSG00000075577, Gm10868, Mnf
Chromosome Location
Chr.5: 142401497 - 142462015 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199068.2 NP_951031.2 2 - 3 737 117
GenBank mRNA AK144418.1 - 2 - 3 630 117
AK147375.1 - 2 - 3 735 117
BC060238.1 - 2 - 3 717 117
L26507.1 - 2 - 3 713 117
U95016.1 - 2 - 3 713 117

View Full Product Details