Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Foxk1
Entrez Gene ID
17425
Gene Name
forkhead box K1
Gene Aliases
A630048H08Rik, AI463295, ENSMUSG00000075577, Gm10868, Mnf
Chromosome Location
Chr.5: 142401497 - 142462015 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199068.2 NP_951031.2 5 - 6 1228 112
GenBank mRNA AK144418.1 - 5 - 6 1121 112
AK147375.1 - 5 - 6 1226 112
BC060238.1 - 5 - 6 1208 112

View Full Product Details