Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Foxk1
Entrez Gene ID
17425
Gene Name
forkhead box K1
Gene Aliases
A630048H08Rik, AI463295, ENSMUSG00000075577, Gm10868, Mnf
Chromosome Location
Chr.5: 142401497 - 142462015 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199068.2 NP_951031.2 6 - 7 1399 51
GenBank mRNA AK144418.1 - 6 - 7 1292 51
AK147375.1 - 6 - 7 1397 51
BC060238.1 - 6 - 7 1379 51
L26507.1 - 6 - 7 1375 51

View Full Product Details