Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc28
Entrez Gene ID
209683
Gene Name
tetratricopeptide repeat domain 28
Gene Aliases
2310015L07Rik, 6030435N04, AI428795, AI851761, BC002262, TPRBK
Chromosome Location
Chr.5: 110879784 - 111289780 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267622.1 NP_001254551.1 22 - 23 5892 107
GenBank mRNA AK031458.1 - 5 - 6 486 107
AK081382.1 - 5 - 6 364 107
AK122427.1 - 15 - 16 4320 107
BC046779.1 - 16 - 17 3556 107
BC060735.1 - 15 - 16 2552 107

View Full Product Details