The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005247.2 | NP_001005247.2 | 11 - 12 | 1826 | 91 |
NM_001005248.2 | NP_001005248.2 | 11 - 12 | 1802 | 91 | |
NM_001167864.1 | NP_001161336.1 | 11 - 12 | 1727 | 91 | |
GenBank mRNA | AK129265.1 | - | 11 - 12 | 1787 | 91 |
AK141088.1 | - | 11 - 12 | 1624 | 91 | |
AK154612.1 | - | 11 - 12 | 1546 | 91 | |
AK157182.1 | - | 11 - 12 | 1593 | 91 | |
AK161039.1 | - | 2 - 3 | 197 | 91 | |
BC046405.1 | - | 6 - 7 | 805 | 91 | |
BC046443.1 | - | 11 - 12 | 1440 | 91 | |
BC082542.1 | - | 11 - 12 | 1727 | 91 |