The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005247.2 | NP_001005247.2 | 22 - 23 | 3821 | 106 |
NM_001005248.2 | NP_001005248.2 | 22 - 23 | 3797 | 106 | |
NM_001167864.1 | NP_001161336.1 | 22 - 23 | 3722 | 106 | |
GenBank mRNA | AF534396.1 | - | 22 - 23 | 3618 | 106 |
AF534397.1 | - | 22 - 23 | 3594 | 106 | |
AF534398.1 | - | 20 - 21 | 3183 | 106 | |
AF534399.1 | - | 23 - 24 | 3749 | 106 | |
AK129265.1 | - | 22 - 23 | 3744 | 106 | |
AK140780.1 | - | 11 - 12 | 2003 | 106 | |
AK141088.1 | - | 21 - 22 | 4447 | 107 | |
AK154612.1 | - | 22 - 23 | 3551 | 106 | |
AK157182.1 | - | 22 - 23 | 3587 | 106 | |
AK161039.1 | - | 13 - 14 | 2192 | 106 | |
BC046405.1 | - | 17 - 18 | 2800 | 106 | |
BC046443.1 | - | 22 - 23 | 3435 | 106 | |
BC082542.1 | - | 22 - 23 | 3722 | 106 |