Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx51
Entrez Gene ID
69663
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 51
Gene Aliases
2310061O04Rik, AA537002, C81583, R75505
Chromosome Location
Chr.5: 110653434 - 110660496 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027156.3 NP_081432.2 14 - 15 1923 74
GenBank mRNA AK010015.1 - 5 - 6 527 74
AK141361.1 - 14 - 15 1904 74
AK189537.1 - 1 - 2 46 74
BC023700.1 - 11 - 12 1186 74
BC060646.1 - 14 - 15 1900 74
BC069876.1 - 7 - 8 751 74

View Full Product Details