The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027156.3 | NP_081432.2 | 1 - 2 | 334 | 70 |
GenBank mRNA | AK141361.1 | - | 1 - 2 | 314 | 70 |
AK152599.1 | - | 1 - 2 | 302 | 70 | |
BC060646.1 | - | 1 - 2 | 311 | 70 |