The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021555.2 | NP_067530.2 | 5 - 6 | 1082 | 70 |
GenBank mRNA | AB041552.1 | - | 5 - 6 | 1082 | 70 |
AK085815.1 | - | 5 - 6 | 1076 | 70 | |
BC061251.1 | - | 5 - 6 | 1046 | 70 |