Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfp41
Entrez Gene ID
22701
Gene Name
zinc finger protein 41
Gene Aliases
CTfin92, Zfp-41
Chromosome Location
Chr.15: 75616142 - 75627446 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001044718.2 NP_001038183.1 1 - 2 139 65
GenBank mRNA BC053927.1 - 1 - 2 64 65
BY254918.1 - 1 - 2 139 65

View Full Product Details