Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gtf2ird2
Entrez Gene ID
114674
Gene Name
GTF2I repeat domain containing 2
Gene Aliases
1700012P16Rik
Chromosome Location
Chr.5: 134183215 - 134218146 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053266.1 NP_444496.1 12 - 13 1166 113
GenBank mRNA AK082488.1 - - 1335 113
AK209051.1 - 2 - 3 152 113
AY014963.1 - 12 - 13 1166 113
AY116023.1 - 12 - 13 1177 113
BC057179.1 - 11 - 12 931 113
BC132157.1 - 12 - 13 1074 113
BC132159.1 - 12 - 13 1074 113

View Full Product Details