The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012001.2 | NP_036131.1 | 9 - 10 | 1260 | 77 |
GenBank mRNA | AF071314.1 | - | 9 - 10 | 1129 | 77 |
AK003752.1 | - | 9 - 10 | 1127 | 77 | |
AK009955.1 | - | 9 - 10 | 1132 | 77 | |
AK167833.1 | - | 9 - 10 | 1146 | 77 | |
BC020461.1 | - | 5 - 6 | 1682 | 77 | |
BC116800.1 | - | 9 - 10 | 1114 | 77 | |
BC116826.1 | - | 9 - 10 | 1114 | 77 | |
CD774670.1 | - | 1 - 2 | 77 |