Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem2
Entrez Gene ID
83921
Gene Name
transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.19: 21777508 - 21858437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033759.2 NP_001028931.1 11 - 12 2639 115
NM_031997.4 NP_114386.3 12 - 13 2721 115
GenBank mRNA AK129352.1 - 6 - 7 940 115
BC019745.1 - 3 - 4 362 115
BC076570.1 - 11 - 12 2245 115
BC089353.1 - 11 - 12 2621 115
BU058172.1 - 2 - 3 153 115
CA752446.1 - 4 - 5 579 115

View Full Product Details