Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem2
Entrez Gene ID
83921
Gene Name
transmembrane protein 2
Gene Aliases
-
Chromosome Location
Chr.19: 21777508 - 21858437 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033759.2 NP_001028931.1 7 - 8 2020 100
NM_031997.4 NP_114386.3 8 - 9 2102 100
GenBank mRNA AK129352.1 - 2 - 3 321 100
BC076570.1 - 7 - 8 1626 100
BC089353.1 - 7 - 8 2002 100
BQ895808.1 - - 322 100

View Full Product Details