Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jak2
Entrez Gene ID
16452
Gene Name
Janus kinase 2
Gene Aliases
Fd17
Chromosome Location
Chr.19: 29251776 - 29313080 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048177.2 NP_001041642.1 10 - 11 1848 125
NM_008413.3 NP_032439.2 11 - 12 1931 125
GenBank mRNA AK087255.1 - 10 - 11 1815 125
BC054807.1 - 10 - 11 1611 125
BC059834.1 - 10 - 11 1611 125
L16956.1 - 10 - 11 1600 125

View Full Product Details