Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Jak2
Entrez Gene ID
16452
Gene Name
Janus kinase 2
Gene Aliases
Fd17
Chromosome Location
Chr.19: 29251776 - 29313080 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001048177.2 NP_001041642.1 14 - 15 2323 118
NM_008413.3 NP_032439.2 15 - 16 2406 118
GenBank mRNA L16956.1 - 14 - 15 2075 118

View Full Product Details