The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009160.2 | NP_033186.1 | 6 - 7 | 740 | 97 |
GenBank mRNA | BC003705.1 | - | 6 - 7 | 691 | 97 |
BG085186.2 | - | 2 - 3 | 179 | 97 | |
L40156.1 | - | 5 - 6 | 668 | 97 |