Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Veph1
Entrez Gene ID
72789
Gene Name
ventricular zone expressed PH domain-containing 1
Gene Aliases
2810471M23Rik, Veph
Chromosome Location
Chr.3: 66053558 - 66297066 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145820.3 NP_665819.2 8 - 9 1719 76
GenBank mRNA AB085187.1 - 8 - 9 1701 76
BC075649.1 - 8 - 9 1719 76
BC128271.1 - 7 - 8 1426 76

View Full Product Details