The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053177.1 | NP_444407.1 | 13 - 14 | 1782 | 70 |
GenBank mRNA | AF302009.1 | - | 13 - 14 | 1779 | 70 |
AK028385.1 | - | 13 - 14 | 1759 | 70 | |
AK034471.1 | - | 13 - 14 | 1793 | 70 | |
AK038594.1 | - | 13 - 14 | 1793 | 70 | |
AK049606.1 | - | 13 - 14 | 1783 | 70 | |
AK195153.1 | - | 2 - 3 | 151 | 70 | |
BC005651.1 | - | 13 - 14 | 1782 | 70 | |
BC020377.1 | - | 9 - 10 | 2224 | 70 |