Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc18a3
Entrez Gene ID
20508
Gene Name
solute carrier family 18 (vesicular monoamine), member 3
Gene Aliases
VAChT, VAT
Chromosome Location
Chr.14: 32462437 - 32464850 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021712.2 NP_068358.2 1 - 1 1964 57
GenBank mRNA AK158592.1 - 1 - 1 1966 57
BC120498.1 - 1 - 1 1581 57
BC137705.1 - 1 - 1 1580 57

View Full Product Details