Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Chat
Entrez Gene ID
12647
Gene Name
choline acetyltransferase
Gene Aliases
B230380D24Rik
Chromosome Location
Chr.14: 32408203 - 32465909 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009891.2 NP_034021.1 9 - 10 1296 60
GenBank mRNA AK046406.1 - 9 - 10 1297 60
AK047708.1 - 8 - 9 1188 60
BC119322.1 - 8 - 9 1058 60
BC120589.1 - 8 - 9 1058 60

View Full Product Details