Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tbc1d14
Entrez Gene ID
100855
Gene Name
TBC1 domain family, member 14
Gene Aliases
2810413P16Rik, AU043625, C86258, D5Ertd110e, mKIAA1322
Chromosome Location
Chr.5: 36490604 - 36586273 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113362.1 NP_001106833.1 13 - 14 1997 148
NM_001113364.1 NP_001106835.1 10 - 11 1111 148
NM_133910.3 NP_598671.3 12 - 13 1816 148
GenBank mRNA AB064521.1 - 10 - 11 974 148
AB093293.1 - 11 - 12 1610 148
AK029986.1 - 13 - 14 1997 148
BC011420.1 - 10 - 11 989 148
BC042515.1 - 12 - 13 1792 148
BC096446.1 - 12 - 13 1808 148

View Full Product Details