Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Thumpd2
Entrez Gene ID
72167
Gene Name
THUMP domain containing 2
Gene Aliases
2810025A12Rik
Chromosome Location
Chr.17: 81026327 - 81065127 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028138.1 NP_082414.1 1 - 2 154 102
GenBank mRNA AK012806.1 - 1 - 2 137 102
BY133372.1 - 1 - 2 154 102

View Full Product Details