Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nr2c1
Entrez Gene ID
22025
Gene Name
nuclear receptor subfamily 2, group C, member 1
Gene Aliases
4831444H07Rik, 80.3, Eenr, TR2, Tr2-11
Chromosome Location
Chr.10: 94147517 - 94197216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011629.3 NP_035759.3 13 - 14 1787 124
GenBank mRNA AK149374.1 - 13 - 14 1702 124
BC090662.1 - 13 - 14 1763 124
BC094580.1 - 13 - 14 1705 124
U28265.1 - 12 - 13 1596 124
Y11436.1 - 12 - 13 2235 124

View Full Product Details