Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
Nr2c1
Entrez Gene ID
22025
Gene Name
nuclear receptor subfamily 2, group C, member 1
Gene Aliases
4831444H07Rik, 80.3, Eenr, TR2, Tr2-11
Chromosome Location
Chr.10: 94147517 - 94197216 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011629.3 NP_035759.3 10 - 10 1385 236
GenBank mRNA AK146891.1 - 10 - 10 1398 236
AK149374.1 - 10 - 10 1300 236
BC090662.1 - 10 - 10 1361 236
BC094580.1 - 10 - 10 1303 236
L26957.1 - 9 - 9 1194 236
U28265.1 - 9 - 9 1194 236
U30482.1 - 9 - 9 1194 236
Y11436.1 - 9 - 9 1833 236

View Full Product Details