Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Oxgr1
Entrez Gene ID
239283
Gene Name
oxoglutarate (alpha-ketoglutarate) receptor 1
Gene Aliases
Cysltr3, Gm299, Gpr80, Gpr99, P2Y15
Chromosome Location
Chr.14: 120019585 - 120042435 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001490.3 NP_001001490.1 1 - 2 152 62
GenBank mRNA AK142510.1 - 1 - 2 153 62

View Full Product Details