The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001984.2 | NP_001001984.2 | 20 - 21 | 4128 | 88 |
GenBank mRNA | AK141779.1 | - | 6 - 7 | 1369 | 88 |
AK173084.1 | - | 9 - 10 | 1946 | 88 | |
BC036264.1 | - | 8 - 9 | 1827 | 88 | |
BC037117.1 | - | 13 - 14 | 2679 | 88 | |
BC057051.1 | - | 7 - 8 | 1501 | 88 | |
BC076576.1 | - | 20 - 21 | 3693 | 88 |