Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Coro6
Entrez Gene ID
216961
Gene Name
coronin 6
Gene Aliases
-
Chromosome Location
Chr.11: 77462321 - 77470484 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_139128.1 NP_624354.1 3 - 4 446 81
NM_139129.1 NP_624355.1 3 - 4 446 81
NM_139130.2 NP_624356.1 3 - 4 446 81
GenBank mRNA AB070894.1 - 3 - 4 446 81
AB070895.1 - 3 - 4 446 81
AB070896.1 - 3 - 4 446 81
AK158179.1 - 4 - 5 638 81
BC141247.1 - 3 - 4 523 81
BC145575.1 - 3 - 4 523 81

View Full Product Details