Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Csnk2a2
Entrez Gene ID
13000
Gene Name
casein kinase 2, alpha prime polypeptide
Gene Aliases
1110035J23Rik, C77789, CK2
Chromosome Location
Chr.8: 95446096 - 95488904 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009974.3 NP_034104.1 5 - 6 801 85
GenBank mRNA AF012251.1 - 5 - 6 526 85
AJ001420.1 - 5 - 6 771 85
AK002927.1 - 5 - 6 809 85
AK076042.1 - 5 - 6 762 85
BC047143.1 - 5 - 6 791 85
BC057862.1 - 5 - 6 461 85

View Full Product Details