Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ssh2
Entrez Gene ID
237860
Gene Name
slingshot homolog 2 (Drosophila)
Gene Aliases
SSH-2, SSH-2L, mSSH-2L
Chromosome Location
Chr.11: 77216169 - 77460220 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291190.1 NP_001278119.1 3 - 4 438 75
NM_177710.4 NP_808378.2 4 - 5 299 75
GenBank mRNA AB099288.1 - 4 - 5 299 75
AK155232.1 - 3 - 4 429 75
AK158449.1 - 3 - 4 439 75
AK170142.1 - 3 - 4 432 75
BC141392.1 - 4 - 5 293 75
BC141393.1 - 4 - 5 293 75

View Full Product Details