Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pck1
Entrez Gene ID
18534
Gene Name
phosphoenolpyruvate carboxykinase 1, cytosolic
Gene Aliases
AI265463, PEPCK, Pck-1
Chromosome Location
Chr.2: 173153073 - 173159254 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011044.2 NP_035174.1 9 - 10 1557 62
GenBank mRNA AK028046.1 - 9 - 10 1558 62
AK030327.1 - 9 - 10 1557 62
AK133496.1 - 9 - 10 1558 62
AK149525.1 - 9 - 10 1558 62
BC037629.1 - 9 - 10 1552 62

View Full Product Details