Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Slc9a8
- Entrez Gene ID
-
77031
- Gene Name
- solute carrier family 9 (sodium/hydrogen exchanger), member 8
- Gene Aliases
-
1200006P13Rik, 6430709P13Rik, AI182282, NHE-8, NHE8
- Chromosome Location
-
Chr.2: 167420056 - 167477000 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK051791.1 |
- |
3 - 4 |
279 |
114 |
View Full Product Details