Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vat1l
Entrez Gene ID
270097
Gene Name
vesicle amine transport protein 1 homolog-like (T. californica)
Gene Aliases
9430073I07, AI427515, mKIAA1576
Chromosome Location
Chr.8: 114205582 - 114374074 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173016.3 NP_766604.2 7 - 8 1144 76
GenBank mRNA AK122527.1 - 7 - 8 1144 76
BC056927.1 - 7 - 8 1154 76

View Full Product Details