Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cd28
Entrez Gene ID
12487
Gene Name
CD28 antigen
Gene Aliases
-
Chromosome Location
Chr.1: 60746388 - 60773359 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007642.4 NP_031668.3 2 - 3 496 82
GenBank mRNA AK088021.1 - 2 - 3 430 82
BC064058.1 - 2 - 3 410 82
M34563.1 - 2 - 3 466 82

View Full Product Details