Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr78
Entrez Gene ID
242584
Gene Name
WD repeat domain 78
Gene Aliases
BC028975
Chromosome Location
Chr.4: 103034757 - 103114299 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146254.4 NP_666366.4 14 - 15 2133 113

View Full Product Details